Learn More
Selleck Chemical LLC Plerixafor (AMD3100) 8HCl 10mg 155148-31-5 JM 3100 8HCl
SDP
Supplier:  Selleck Chemical LLC S301310MG
Encompass_Preferred
Plerixafor (AMD3100, JM 3100) 8HCl is the hydrochloride of Plerixafor, a chemokine receptor antagonist for CXCR4 and CXCL12-mediated chemotaxis with IC50 of 44 nM and 5.7 nM in cell-free assays, respectively. Plerixafor can be used as an anti-HIV agent. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1103425