Learn More
Apexbio Technology LLC Plerixafor 8HCl (AMD3100 8HCl), 50mg. Cas: 155148-31-5 MFCD: MFCD04974488. CXCR4 antagonist
SDP
Supplier:  Apexbio Technology LLC B146550
Encompass_Preferred
Plerixafor 8HCl (AMD3100 8HCl) is a potent and selective antagonist of CXCL12-mediated chemotaxis and G-protein coupled chemokine receptor (CXCR4) with IC50 values of 5.7 and 44 nM, respectively [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5241