Learn More
Apexbio Technology LLC Plerixafor 8HCl (AMD3100 8HCl), 155148-31-5, MFCD04974488, 25 mg
SDP
Supplier:  Apexbio Technology LLC B146525
Encompass_Preferred
MF: C28H62Cl8N8, MW: 794.47, Solubility: 155.4mg/mL in H2O,insoluble in DMSO. Plerixafor 8HCl (AMD3100 8HCl) is a potent and selective antagonist of CXCL12-mediated chemotaxis and G-protein coupled chemokine receptor (CXCR4) with IC50 values of 5.7 and 44 nM, respectively. Plerixafor 8HCl has shown a high selectivity for CXCR4 compared other chemokine receptors including LTB4 CCR1, CCR2b, CCR4, CCR5, CCR7, CXCR3, etc.
Catalog No. 50-101-2466