Learn More
Selleck Chemical LLC Plerixafor (AMD3100) 10mg 110078-46-1 JM 3100
SDP
Supplier:  Selleck Chemical LLC S803010MG
Encompass_Preferred
Plerixafor (AMD3100, JM 3100) is a chemokine receptor antagonist for CXCR4 and CXCL12-mediated chemotaxis with IC50 of 44 nM and 5.7 nM in cell-free assays, respectively. Plerixafor inhibits human immunodeficiency virus (HIV) replication. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5582