Learn More
Selleck Chemical LLC Plerixafor
SDP
Supplier:  Selleck Chemical LLC S803050MG
Encompass_Preferred
Plerixafor (AMD3100 JM 3100 SID791) is a chemokine receptor antagonist for CXCR4 and CXCL12-mediated chemotaxis with IC50 of 44 nM and 5 7 nM in cell-free assays respectively Plerixafor inhibits human immunodeficiency virus (HIV) replication
Catalog No. 50-136-5583