Learn More
Apexbio Technology LLC Plerixafor (AMD3100), 50mg. Cas: 110078-46-1 MFCD: MFCD05662218. CXCR4 chemokine receptor antagonist
SDP
Supplier:  Apexbio Technology LLC A202550
Encompass_Preferred
Plerixafor (AMD3100) is a small-molecule antagonist of CXCR4 and CXCL12-mediated chemotaxis with IC50 of 44 nM and 5.7 nM, respectively. CXCR4 and SDF-1 are key factors in regulating cancer cell invasion and metastasis, and plerixafor can prevent the binding of SDF-1 to CXCR4 for inhibiting cancer metastasis. Plerixafor interfered with CXCL12/CXCR4 mediated retention of hematopoietic stem cells in the bone marrow, and resulted in their mobilization to the blood. Plerixafor amplified the release of circulating neutrophils from the oriented area in the lung, while simultaneously preventing neutrophil returned to the bone marrow. Three adults with WHIM syndrome were subcutaneously injected 0.01 to 0.02 mg/kg plerixafor twice daily for 6 months, circulating leukocytes were constantly increased, and associated with fewer infections. Other sizes are available. Please inqury us for quote.
Catalog No. NC1251956