Learn More
Medchemexpress LLC HY-50912 50mg , Plerixafor (octahydrochloride) CAS:155148-31-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY5091250MG
Encompass_Preferred
HY-50912 50mg Plerixafor (octahydrochloride) CAS: 155148-31-5 Plerixafor octahydrochloride (AMD3100 octahydrochloride) is a selective CXCR4 antagonist with an IC50 of 44 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0261