Learn More
Medchemexpress LLC Plerixafor (octahydrochloride) | 155148-31-5 | 99.1% | C28H54N8.8HCl | 5 MG
SDP
Supplier:  Medchemexpress LLC HY509125MG
Encompass_Preferred
Plerixafor octahydrochloride is a selective CXCR4 antagonist with an IC50 of 44 nM. It is intended for laboratory research and substance manufacturing, not for patient use.
  • Selective CXCR4 antagonist
  • Potent inhibitor of CXCL12-mediated chemotaxis (IC50, 5.7 nM)
  • Inhibits CXCL12-mediated TEM in MOLT-4 and U937 cells
  • Reduces perivascular and interstitial fibrosis in vivo
  • Cited in numerous top publications
Catalog No. 50-003-57679