Learn More
Medchemexpress LLC Plerixafor octahydrochloride | 155148-31-5 | MFCD04974488 | 99.1% | 794.47 g/mol | C28H62Cl8N8 | 10 MG

Supplier: Medchemexpress LLC HY5091210MG
Plerixafor octahydrochloride is a small-molecule selective antagonist of the CXCR4 chemokine receptor provided for laboratory research use. It exhibits reported CXCR4 inhibitory activity and is used in studies of stem cell mobilization, HIV, cancer, and immunology.
- Selective CXCR4 antagonist with IC50 44 nM.
- Chemical formula C28H62Cl8N8 and molecular weight 794.47 g/mol.
- High purity (≈99.1%) suitable for analytical and research applications.
- CAS number 155148-31-5 for unambiguous substance identification.
- Supplied in milligram-scale packages for laboratory research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.