Learn More
TARGETMOL CHEMICALS INC Plerixafor octahydrochloride 2
SDP
Supplier:  TARGETMOL CHEMICALS INC T1776L25MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Plerixafor octahydrochloride (JM3100 octahydrochloride) blocks the binding of stromal cell-derived factor (SDF-1alpha) to the cellular receptor CXCR4, resulting in hematopoietic stem cell (HSC) release from bone marrow and HSC movement into the peripheral circulation. Plerixafor is a bicyclam with hematopoietic stem cell-mobilizing activity. Purity 99.96%
Catalog No. 50-287-8548