Learn More
Sigma Aldrich Fine Chemicals Biosciences PLGA nanoparticles 100 nm50MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 805092
Encompass
The particle size of the PLGA nanoparticles are closely dependent on both the nature and concentration of the stabilizer and polymer.
Catalog No. 50-290-7257