Learn More
Medchemexpress LLC Plga-peg-mal (60kda-3.4kda, la:ga ratio 75:25) | 95.0% | 60kDA-3.4kDA | 50 MG

Supplier: Medchemexpress LLC HY14877550MG
PLGA-PEG-MAL (60kDA-3.4kDA, LA:GA ratio 75:25) is a poly(lactide-co-glycolide)-block-poly(ethylene glycol) (PLGA-PEG-Mal) nanoparticle. It has a molecular weight range of 60kDA to 3.4kDA and a 75:25 lactic acid (LA) to glycolic acid (GA) ratio. This product is for research use only.
- Type: poly(lactide-co-glycolide)-block-poly(ethylene glycol) nanoparticles
- Molecular weight: 60kDA to 3.4kDA
- Lactic acid to glycolic acid ratio: 75:25
- Purity: 95.0%
- Appearance: solid
- Color: white to off-white
- Storage: powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month
- Solubility in DMSO: 100 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.