Learn More
Medchemexpress LLC PLGA12000-PEG2000-NH2 | 98.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14539650MG
Encompass_Preferred
PLGA-PEG-NH2 is a material used to synthesize nanomicelles. Its nanomicelles serve as an efficient delivery system for Irinotecan, targeting colorectal cancer and hepatocellular carcinoma.
  • Used to synthesize nanomicelles
  • Efficient delivery system for Irinotecan
  • Targets colorectal cancer and hepatocellular carcinoma
  • Related to oligonucleotides, polymers, biochemical assay reagents, and drug delivery
  • Applicable in cancer, cancer targeted therapy, and cancer metabolism and metastasis
Catalog No. 50-003-55275