Learn More
Alkali Scientific Plicamycin [Mithramycin A], 1 Mg
SDP
Supplier:  Alkali Scientific PLI001CP
Encompass_Preferred
Plicamycin (Mithramycin A), provided in a 1mg powder form, is an anticancer antibiotic with a broad range of applications in both clinical and laboratory settings. This powerful agent is used in the treatment of various cancers, including testicular cancer and osteosarcoma, due to its ability to inhibit DNA synthesis and cell division. In research, plicamycin is employed to study gene expression regulation and as an inhibitor in molecular biology experiments. The 1mg powder format allows for flexible dosing, making it ideal for experimental protocols where precise amounts are needed. It also exhibits activity against various types of microbial infections, aiding in both oncology and antimicrobial studies.
Catalog No. 50-006-61544