Learn More
Medchemexpress LLC HY-14444 10mg , Plinabulin CAS:714272-27-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1444410MG
Encompass_Preferred
HY-14444 10mg Plinabulin CAS: 714272-27-2 Plinabulin (NPI-2358) is a vascular disrupting agen (VDA) against tubulin-depolymerizing with an IC50 of 9.8 nM against HT-29 cells. Plinabulin binds the colchicine binding site of β-tubulin preventing polymerization and has potent inhibitory to tumor cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0857