Learn More
Selleck Chemical LLC Plixorafenib
SDP
Supplier:  Selleck Chemical LLC S79651G
Encompass_Preferred
Plixorafenib (PLX8394) is a next-generation orally available small-molecule BRAF inhibitor with IC50 values of 3 8 nM 14 nM and 23 nM for BRAF(V600E) WT BRAF and CRAF respectively It has potential antineoplastic activity
Catalog No. 50-000-01258