Learn More
Medchemexpress LLC Plk4-in-1 | 1247001-12-2 | 99.8% | 431.23 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY134775
Encompass_Preferred
Plk4-in-1 is a Plk4 inhibitor with an IC50 of ≤ 0.1 μM. It is for research use only.
  • Plk4 inhibitor
  • IC50 of ≤ 0.1 μM (Plk4)
  • High purity (99.84%)
Catalog No. 50-003-64278