Learn More
Cayman Chemical Plnt Mulberry leaf Phytol 50g
SDP
Supplier:  Cayman Chemical 1740150 G
Encompass_Preferred
A degradation product of chlorophyll that is converted to phytanic acid, which can accumulate to toxic levels in a number of metabolic disorders; activates RXRs (Kis range from 2.3-67.2 µM) and promotes the activity of PPAR/RXR heterodimers
Catalog No. 50-296-5314