Learn More
Cayman Chemical PlntamajosIde 5mg
SDP
Supplier:  Cayman Chemical 272965 MG
Encompass_Preferred
A phenylethanoid with diverse biological activities; scavenges DPPH radicals in a cell-free assay (IC50 = 11.8 µM); inhibits 5- and 15-LO (IC50s = 0.375 and 96 µM, respectively); decreases viability of TE-1 ESCC cells at 20-320 UG/ml, as well as inhibits LPS-induced EMT in Eca-109 and TE-1 cells and decreases phosphorylated NF-κB levels in TE-1 cells; inhibits arachidonic acid-induced mouse EAr edema at 3 mg/ear
Catalog No. 50-293-9047