Learn More
Medchemexpress LLC PLP (180-199) TFA | 2085.34 | 10 MG
Supplier: Medchemexpress LLC HYP5462A10MG
'''PLP (180-199) TFA is the trifluoroacetic acid (TFA) salt form of PLP (180-199), a biologically active peptide. It is an amino acids 180-199 fragment of myelin proteolipid protein (PLP). PLP is the most abundant myelin protein of the central nervous system and has been utilized in studies of multiple sclerosis (MS). This peptide induces the activation and differentiation of T cells, and also induces experimental autoimmune encephalomyelitis (EAE) in mouse models.
- Trifluoroacetic acid (TFA) salt form
- Induces activation and differentiation of T cells
- Induces experimental autoimmune encephalomyelitis (EAE) in mouse models
- Amino acid fragment (180-199) of myelin proteolipid protein
- Utilized in multiple sclerosis (MS) studies