Learn More
ABclonal Technology PLP1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A14251500UL
Encompass_Preferred
This gene encodes a transmembrane proteolipid protein that is the predominant component of myelin. The encoded protein may play a role in the compaction, stabilization, and maintenance of myelin sheaths, as well as in oligodendrocyte development and axonal survival. Mutations in this gene cause Pelizaeus-Merzbacher disease and spastic paraplegia type 2. Alternatively splicing results in multiple transcript variants, including the DM20 splice variant.
Catalog No. 50-001-08975