Learn More
MSC Made in USA Pallet Cover Liner, 48x42x48in, 1 mil, 150/Pack
Supplier:  MSC 39593827
Encompass
These large, gusseted poly bags are designed for use as pallet covers and bin liners to protect equipment and pallets from dirt and dust. They can also line large cartons to protect products from moisture.
  • Pre-cut and perforated on a roll for easy use
  • Clear poly construction allows for visibility of pallet content
  • Overall width of 48 in
  • Overall length of 42 in
  • Overall thickness of 1 mil
  • Protects equipment and pallets from dirt and dust
  • Lines large cartons to protect products from moisture
Catalog No. 50-191-9765