Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Pallet Cover Liner, 58 x 43 x 76 in, 50/Case

Supplier:  MSC 39593942

Encompass

These large, gusseted poly bags are designed to cover equipment and pallets, protecting large items from dirt and dust. They can also be used to line large cartons, shielding products from moisture. The clear poly material allows for easy visibility of the pallet contents.

  • Large, gusseted poly bags
  • Protects equipment and pallets from dirt and dust
  • Shields products from moisture when lining cartons
  • Pre-cut and perforated on a roll for easy use
  • Clear poly for content visibility
  • Dimensions: 58 in Wide x 43 in Long x 76 in High
  • Features a 2 mil thickness

Catalog No. 50-191-9776


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.