Learn More
Beckman Coulter Hematology, Coulter AcT 5diff OV Hematology Analyzer with Autoloader
Supplier:  Beckman Coulter 361656
Encompass
The Beckman Coulter Microfuge 18 Centrifuge is a compact benchtop microcentrifuge designed for quick spin-downs and separations. It has a maximum speed of 13,300 rpm and maximum RCF of 16,060 xg. The rotor accommodates 18 x 1.5/2.0mL microcentrifuge tubes. It features a compact footprint, digital display, 10 acceleration/deceleration rates, and a pulse key for short spin cycles. The Microfuge 18 is ideal for DNA/RNA extractions, quick spin-downs, protein precipitations, and other molecular biology applications requiring microcentrifugation up to 16,060 xg.
Catalog No. NC1898075