Learn More
MSC Balax Thread Plug Gage, 7/16-20, 3B Class, No Go, 1 ea
Supplier:  MSC 05383229
Encompass
This thread plug gage is designed for inspecting internal threads, specifically for 7/16-20 size with a 3B class fit. Constructed from high speed tool steel, the gage provides high durability and accurate thread measurements for industrial quality control applications.
  • Features a 3B class fit for precise thread inspection
  • Constructed from high speed tool steel for wear resistance
  • Hardness rating of 63-65 Rockwell
  • Bright finish coating
  • Equipped with a #2 taperlock handle size
  • Designed with a 3/8 in. thread length and 1-3/8 in. overall length
  • Includes 2.5 chamfer
Catalog No. 50-005-12708