Learn More
MSC Balax Thread Plug Gage, No-Go Member, 5/16-18, 2B, 1 ea
Supplier:  MSC 05381199
Encompass
This NO GO thread gage member is designed for verifying the internal thread fit of 5/16-18 2B threaded components. Constructed from high-speed tool steel, this gage provides the durability required for precision measurement tasks.
  • Hardened to 63-65 Rockwell for extended wear resistance
  • Features a bright finish
  • Compatible with #1 Taperlock handles
  • Designed as a NO GO member for accurate class 2B inspection
  • Thread length of 0.3125 in. (5/16 in.)
  • Overall length of 1.3125 in. (1-5/16 in.)
Catalog No. 50-005-12709