Learn More
Waters Corp Waters Corporation Plunger Seal Kit, For 1525u Pumps, HPLC, 2/package
Supplier:  Waters Corp 205000202
Encompass_Preferred
This plunger seal kit is a replacement part for 1525u pumps. It includes two bearings and two plunger seals. Note that seal removal and insertion tools are not included with this kit.
  • Designed for HPLC systems.
  • Functions as a rebuild kit.
  • Replacement schedule is as required.
  • Contains two bearings.
  • Contains two plunger seals.
Catalog No. 50-791-644