Learn More
Bio X Cell InVivoPlus anti-mouse CD20, Clone: MB20-11, Size: 5 mg
Supplier:  Bio X Cell BP0356R005MG
Encompass_Preferred
InVivoPlus anti-mouse CD20, Clone: MB20-11, Size: 5 mg, Isotype: Mouse IgG2c, κ, Applications: in vivo B cell depletion, Western blot
Catalog No. 50-217-2986