Learn More
Abcam (±)-1,2-Didecanoylglycerol (10:0), Protein kinase C activator, 50MG
Supplier:  Abcam AB14520650MG
Encompass_Preferred
MW 400.6 g/mol, Purity >98%. Non-physiologic diacylglycerol (DAG). Cell-permeable protein kinase C activator in platelets. Enhances anterior pituitary hormone secretion in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5397