Learn More
Abcam (±)-Bisoprolol hemifumarate, beta1 antagonist, 10MG
Supplier:  Abcam AB12078310MG
Encompass_Preferred
MW 383.48 g/mol, Purity >99%. Selective β₁ antagonist (Ki = 22.4 nM). Antihypertensive agent.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7277