Learn More
Abcam (±)-SKF38393 hydrochloride, D1-like dopamine receptor agonist, 100MG
Supplier:  Abcam AB120740100MG
Encompass_Preferred
MW 291.77 Da, Purity >98%. D1-like dopamine receptor agonist. pKi values are 9 (D1), 6.8 (D2), 5.3 (D3), 6.0 (D4) and 9.3 (D5). Ki values are 151 and 2364 nM for D1-like and D2-like dopamine receptors, respectively. Centrally active upon systemic administration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7315