Learn More
ABclonal Technology PLVAP Rabbit pAb
SDP
Supplier:  ABclonal Technology A15906500UL
Encompass_Preferred
Predicted to enable identical protein binding activity. Involved in MAPK cascade, positive regulation of cellular extravasation, and tumor necrosis factor-mediated signaling pathway. Located in cell surface. Colocalizes with caveola. Implicated in congenital diarrhea.
Catalog No. 50-001-07516