Learn More
Medchemexpress LLC HY-114153 10mg Medchemexpress, PLX5622 CAS:1303420-67-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11415310MG
Encompass_Preferred
Medchemexpress, HY-114153 10mg PLX5622 CAS:1303420-67-8 PLX5622 is a highly selective brain penetrant and oral active CSF1R inhibitor with an Ki of 5.9 nM, for extended and specific microglial elimination, preceding and during pathology development. PLX5622 demonstrates desirable PK properties in varies animals. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9289