Learn More
Medchemexpress LLC HY-114153A 50mg , PLX5622 (hemifumarate) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY114153A50MG
Encompass_Preferred
HY-114153A 50mg PLX5622 (hemifumarate) CAS: PLX5622 hemifumarate is a highly selective brain penetrant and orally active CSF1R inhibitor (IC50=0.016 µM; Ki=5.9 nM). PLX5622 hemifumarate allows for extended and specific microglial elimination, preceding and during pathology development. PLX5622 hemifumarate demonstrates desirable PK properties in varies animals. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0561