Learn More
Selleck Chemical LLC PMA
SDP
Supplier:  Selleck Chemical LLC S779125MG
Encompass_Preferred
PMA (Phorbol 12-myristate 13-acetate) a potent activator of PKC is active at nanomolar concentrations Phorbol 12-myristate 13-acetate (PMA) induces sphingosine-1-phosphate (S1P) PMA has strong excitability on the skin and mucous membrane Special protection is required when using PMA Wear gloves and mask to avoid direct contact in any way
Catalog No. 50-000-00583