Learn More
Electron Microscopy Sciences PMFS (Phenylmethylsulfonyl Fluoride) 1 GR

Supplier: Electron Microscopy Sciences 19330
CAS #329-98-6 C₆H₅CH₂SO₂F F.W. 174.20
Assay: >99.0%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.