Learn More
Enzo Life Sciences 2-(Phosphonomethyl)-pentanedioic acid (5mg). CAS: 173039-10-6
SDP
Supplier:  Enzo Life Sciences ALX550358M005
Encompass_Preferred
Potent inhibitor of the neuropeptidase N-acetylated α-linked acidic dipeptidase (NAALADase), which is a membrane-bound peptidase that hydrolyses N-acetyl-L-aspartate-L-glutamate (NAAG) (ALX-151-020), a major brain peptide. Alternative name: PMPA. Purity: ≥97% (HPLC). Solubility: Soluble in water, methanol or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-9543