Learn More
ABclonal Technology PMPCB Rabbit pAb
SDP
Supplier:  ABclonal Technology A125491000UL
Encompass_Preferred
This gene is a member of the peptidase M16 family and encodes a protein with a zinc-binding motif. This protein is located in the mitochondrial matrix and catalyzes the cleavage of the leader peptides of precursor proteins newly imported into the mitochondria, though it only functions as part of a heterodimeric complex.
Catalog No. 50-001-14114