Learn More
Apexbio Technology LLC PMSF 329-98-6 10g
SDP
Supplier:  Apexbio Technology LLC A258710000
Encompass_Preferred
Phenylmethanesulfonyl fluoride (PMSF CAS 329-98-6) is an irreversible serine protease inhibitor widely utilized in biomedical research It covalently modifies serine residues within the active sites of targeted enzymes effectively inhibiting their catalytic function PMSF is commonly used to inhibit neuropathy target esterase (NTE) a protective enzyme involved in delayed neurotoxicity induced by organophosphates Its inhibition of NTE significantly enhances susceptibility to organophosphate-induced delayed neuropathy (OPIDP) Additionally PMSF specifically targets -glutamyl transpeptidase serving as an enzymatic site-directed inactivator to probe serine enzyme mechanisms
Catalog No. 50-000-63558