Learn More
Carbosynth LLC. Phenylmethylsulfonyl fluoride, 329-98-6, MFCD00007424, 10 g
SDP
Supplier:  Carbosynth LLC. FP1225710G
Encompass
Formula: C7H7FO2S, Molecular weight: 174.20 g/mol, 99%, Store at room temperature, Synonyms: Phenylmethanesulfonyl fluoride; alpha-Toluenesulfonyl fluoride.
Catalog No. 50-121-7868