Learn More
Selleck Chemical LLC PMSF 10mM/1mL 329-98-6 Phenylmethylsulfonyl Fluoride
SDP
Supplier:  Selleck Chemical LLC S302510MM/1ML
Encompass_Preferred
PMSF (Phenylmethylsulfonyl Fluoride) is an irreversible serine/cysteine protease inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3274