Learn More
Gold Biotechnology Inc PMSF
SDP
Supplier:  Gold Biotechnology Inc P470100
Encompass_Preferred
PMSF is a serine protease inhibitor that irreversibly inhibits serine proteases by sulfonylation of the serine residue in the active site of the protease It is widely used in protocols for the isolation of proteins
Catalog No. 50-005-62153