Learn More
Alkali Scientific PMSF [Phenylmethylsulfonyl fluoride], 1 G
SDP
Supplier:  Alkali Scientific PMS001CP
Encompass_Preferred
PMSF (Phenylmethylsulfonyl fluoride) is a potent protease inhibitor commonly used in protein protection protocols. This 1g quantity is ideal for laboratories where smaller quantities are needed for specific protein extraction or purification experiments. PMSF inhibits serine and cysteine proteases by irreversibly binding to their active sites, preventing unwanted protein degradation during sample preparation. It is widely used in cell biology and biochemistry for the isolation of intact proteins, maintaining their structure and function. PMSF is an essential reagent in experiments involving enzyme activity assays, proteomics studies, and tissue lysate preparation, helping preserve protein integrity during experimental procedures.
Catalog No. 50-006-61193