Learn More
Alkali Scientific PMSF [Phenylmethylsulfonyl fluoride], 5 G
SDP
Supplier:  Alkali Scientific PMS005CP
Encompass_Preferred
PMSF (Phenylmethylsulfonyl fluoride) is a potent protease inhibitor that specifically targets serine proteases, effectively preventing protein degradation during biochemical and cellular analysis. Available in powder form, PMSF is used in research and laboratory applications that require the inhibition of unwanted protease activity. It is commonly employed in protein extraction procedures to stabilize proteins during isolation and purification, preventing proteolysis. PMSF is particularly useful in studies related to metabolic pathways, enzyme function, and protein synthesis. This reagent is essential in maintaining protein integrity in both in vitro and in vivo experiments where protease inhibition is crucial for accurate results.
Catalog No. 50-006-61202