Learn More
Abcam PMSF, protease inhibitor, 10G
Supplier:  Abcam AB14103210G
Encompass_Preferred
MW 174.19 Da, Purity >98%. Non-specific protease inhibitor; binds key active site serine residue in serine proteases, acetylcholinesterases, and thiol proteases. Suitable for use in vitro; in the preparation of cell lysates, and in vivo where it has been shown to block anandamide (ab120429) metabolism. Able to rapidly cross the BBB.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3376