Learn More
Medchemexpress LLC PMX 205 | 514814-49-4 | 99.5% | 839.04 | 50 MG
Supplier: Medchemexpress LLC HY11013650MG
PMX 205 is a potent complement C5a receptor (C5aR; CD88) antagonist. It is orally active and selective.
- Potent complement C5a receptor (C5aR; CD88) antagonist
- Orally active and selective C5aR antagonist
- Extended survival time and reduced end-stage motor scores in a rat model of amyotrophic lateral sclerosis
- Reduced astroglial proliferation in the lumbar spinal cord in a rat model of amyotrophic lateral sclerosis
- Decreased fibrillar plaque load in Tg2576 mice and significantly reduced hyperphosphorylated tau in 3xTg mice in models of Alzheimer's disease