Learn More
Medchemexpress LLC PND-1186 (VS-4718) | 1061353-68-1 | 99.9% | 501.50 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13917100MG
Encompass_Preferred
PND-1186 (VS-4718) is a potent, highly-specific, and reversible inhibitor of FAK with an IC50 of 1.5 nM. It selectively promotes tumor cell apoptosis.
  • Potent, highly-specific, and reversible FAK inhibitor.
  • Selectively promotes tumor cell apoptosis.
  • Inhibits FAK Tyr-397 phosphorylation in breast carcinoma cells.
  • Reduces tumor growth by inducing apoptosis.
Catalog No. 50-003-48226