Learn More
Grainger Pneumatic Vibrator 75 lb 8000 vpm
Supplier:  Grainger 14K261
Encompass_Preferred
The VIBCO Pneumatic Vibrator is a 75 lbf force vibrator that operates on compressed air. It has a frequency of 3600 vpm and is made of cast iron with a powder coated finish. The vibrator has a 3/4" NPT air inlet and comes with a 6 foot air hose. It is designed for material flow assistance and hopper or chute cleaning. The vibrator measures 9" x 5" x 6" and weighs 22 lbs. It requires 60-100 psi of compressed air to operate and has an air consumption of 25 cfm. The vibrator can be mounted in any orientation using the four mounting holes.
Catalog No. 50-275-0371