Learn More
Medchemexpress LLC Pneumocandin B0 | 135575-42-7 | 96.5% | 1065.21 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY17578100MG
Encompass_Preferred
Pneumocandin B0 is the precursor of the synthetic antifungal agent Cancidas. It can be isolated from the fungus Glarea lozoyensis and serves as an intermediate for the synthesis of the echinocandin antifungal agent, which inhibits the synthesis of β-(1,3)-D-glucan in the fungal cell wall. It shows inhibitory activity against invasive aspergillosis and Candida infections.
  • Precursor of synthetic antifungal agent
  • Isolated from Glarea lozoyensis fungus
  • Intermediate for antifungal agent synthesis
  • Inhibits β-(1,3)-D-glucan synthesis in fungal cell wall
  • Exhibits inhibitory activity against invasive aspergillosis
  • Exhibits inhibitory activity against Candida infections
Catalog No. 50-003-87504