Learn More
Medchemexpress LLC Pneumocandin B0 | 135575-42-7 | 96.5% | 1065.21 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY17578
Encompass_Preferred
Pneumocandin B0 (L-688786) is a precursor to the synthetic antifungal agent Cancidas (Caspofungin acetate). It can be isolated from the fungus *Glarea lozoyensis*. It serves as an intermediate for the synthesis of Cancidas, an echinocandin antifungal agent that inhibits β-(1,3)-D-glucan synthesis in fungal cell walls, demonstrating inhibitory activity against invasive aspergillosis and Candida infections. This compound is for research use only.
  • Isolated from the fungus Glarea lozoyensis
  • Serves as an intermediate for antifungal agent synthesis
  • Inhibits β-(1,3)-D-glucan synthesis in fungal cell walls
  • Shows inhibitory activity against invasive aspergillosis and Candida infections
Catalog No. 50-003-83942